siRNA Details
| Gene Symbol: Itpripl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033380.3 | NP_001028552.1 | 1 | 1763 | ||
| GenBank mRNA | AK134593.1 | - | 1 | 1711 | ||
| AK137678.1 | - | 1 | 1765 | |||
| AK149729.1 | - | 1 | 1677 | |||
| BC150856.1 | - | 1 | 1414 | 9087767 | ||
| BC150864.1 | - | 1 | 1414 | 9087775 | ||

Itpripl2
Genomic Map