siRNA Details
Gene Symbol: Fsd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195284.1 | NP_001182213.1 | 4 | 432 | ||
NM_176966.4 | NP_795940.1 | 4 | 432 | |||
GenBank mRNA | AK038893.1 | - | 4 | 410 | ||
AK052914.1 | - | 4 | 475 | |||
AK140020.1 | - | 4 | 416 | |||
BB662298.1 | - | 4 | 470 | |||
BC130265.1 | - | 4 | 348 | 40142990 |
