siRNA Details
| Gene Symbol: Ablim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164491.1 | NP_001157963.1 | 12, 13 | 1344 | ||
| NM_198649.3 | NP_941051.2 | 11, 12 | 1663 | |||
| GenBank mRNA | AK086538.1 | - | 11, 12 | 1443 | ||
| AK173040.1 | - | 11, 12 | 1221 | |||
| BC060275.1 | - | 12, 13 | 1315 | 6822626 | ||
| BC094229.1 | - | 11, 12 | 1637 | 30534236 | ||

Ablim3
Genomic Map