siRNA Details
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177006.3 | NP_795980.2 | 6 | 2294 | ||
GenBank mRNA | AB093290.1 | - | 6 | 1163 | ||
AK034089.1 | - | 7 | 2182 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177006.3 | NP_795980.2 | 6 | 2294 | ||
GenBank mRNA | AB093290.1 | - | 6 | 1163 | ||
AK034089.1 | - | 7 | 2182 |