siRNA Details
| Gene Symbol: Nwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177006.3 | NP_795980.2 | 7 | 4888 | ||
| GenBank mRNA | AB093290.1 | - | 7 | 3756 | ||
| AK136869.1 | - | 1 | 687 | |||
| BC062655.1 | - | 1 | 2378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nwd2| Gene Symbol: Nwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177006.3 | NP_795980.2 | 7 | 4888 | ||
| GenBank mRNA | AB093290.1 | - | 7 | 3756 | ||
| AK136869.1 | - | 1 | 687 | |||
| BC062655.1 | - | 1 | 2378 | |||