siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 6, 7 | 1205 | ||
| NM_177025.5 | NP_795999.2 | 6, 7 | 1205 | |||
| GenBank mRNA | AK037010.1 | - | 7, 8 | 1196 | ||
| AK052533.1 | - | 6, 7 | 1154 | |||
| AK144943.1 | - | 7, 8 | 1249 | |||
| AK164624.1 | - | 7, 8 | 1135 | |||
| AK173076.1 | - | 6, 7 | 1036 | |||
| AY308746.1 | - | 6, 7 | 1205 | |||
| BC067007.1 | - | 6, 7 | 1202 | 5702930 | ||

Cobll1
Genomic Map