siRNA Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027225.1 | NP_081501.1 | 4 | 669 | ||
| NM_177025.5 | NP_795999.2 | 4 | 669 | |||
| GenBank mRNA | AK037010.1 | - | 5 | 660 | ||
| AK052533.1 | - | 4 | 618 | |||
| AK144943.1 | - | 5 | 713 | |||
| AK164624.1 | - | 5 | 599 | |||
| AK173076.1 | - | 4 | 500 | |||
| AY308746.1 | - | 4 | 669 | |||
| BC067007.1 | - | 4 | 666 | 5702930 | ||

Cobll1
Genomic Map