siRNA Details
| Gene Symbol: Auts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177047.3 | NP_796021.2 | 14 | 2497 | ||
| GenBank mRNA | AK147478.1 | - | 3496 | |||
| AK164158.1 | - | 10 | 1937 | |||
| BC058110.1 | - | 2 | 73 | |||
| BC066072.1 | - | 9 | 2270 | |||
| BC096021.1 | - | 14 | 2496 | |||

Auts2
Genomic Map