siRNA Details
| Gene Symbol: Tmem198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177056.4 | NP_796030.1 | 2, 3 | 549 | ||
| GenBank mRNA | AK038957.1 | - | 2, 3 | 510 | ||
| AK082334.1 | - | 1, 2 | 809 | |||
| AK158863.1 | - | 2, 3 | 1023 | |||
| BC059868.1 | - | 2, 3 | 514 | 6809137 | ||

Tmem198
Genomic Map