siRNA Details
| Gene Symbol: Fbxl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199632.1 | NP_001186561.1 | 1, 2 | 140 | ||
| NM_177076.4 | NP_796050.2 | 1, 2 | 140 | |||
| GenBank mRNA | AK029566.1 | - | 1, 2 | 142 | ||
| AK030172.1 | - | 1, 2 | 123 | |||
| BB614373.1 | - | 1, 2 | 141 | |||
| BC145291.1 | - | 1, 2 | 66 | 9053845 | ||
| BC145292.1 | - | 1, 2 | 67 | 9053846 | ||

Fbxl13
Genomic Map