siRNA Details
| Gene Symbol: Sp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177082.4 | NP_796056.2 | 3 | 1138 | ||
| GenBank mRNA | AJ844914.1 | - | 1 | 825 | ||
| AK030745.1 | - | 2 | 957 | |||
| AK053093.1 | - | 3 | 1138 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp8| Gene Symbol: Sp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177082.4 | NP_796056.2 | 3 | 1138 | ||
| GenBank mRNA | AJ844914.1 | - | 1 | 825 | ||
| AK030745.1 | - | 2 | 957 | |||
| AK053093.1 | - | 3 | 1138 | |||