siRNA Details
| Gene Symbol: Fam179a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177087.4 | NP_796061.2 | 4 | 572 | ||
| GenBank mRNA | AK076284.1 | - | 3 | 226 | ||
| AK158647.1 | - | 4 | 357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam179a| Gene Symbol: Fam179a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177087.4 | NP_796061.2 | 4 | 572 | ||
| GenBank mRNA | AK076284.1 | - | 3 | 226 | ||
| AK158647.1 | - | 4 | 357 | |||