siRNA Details
| Gene Symbol: Fam19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182808.3 | NP_877960.1 | 3 | 632 | ||
| GenBank mRNA | AK049880.1 | - | 2 | 523 | ||
| AK050001.1 | - | 3 | 633 | |||
| AY325120.1 | - | 3 | 433 | |||
| BC132179.1 | - | 3 | 467 | 40130456 | ||
| BC132181.1 | - | 3 | 467 | 40130458 | ||

Fam19a1
Genomic Map