siRNA Details
| Gene Symbol: C920006O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036298.1 | - | 1 | 765 | ||
| AK050586.1 | - | 2 | 230 | |||
| AK155758.1 | - | 766 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C920006O11Rik| Gene Symbol: C920006O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036298.1 | - | 1 | 765 | ||
| AK050586.1 | - | 2 | 230 | |||
| AK155758.1 | - | 766 | ||||