siRNA Details
| Gene Symbol: Ric3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038624.1 | NP_001033713.1 | 2 | 256 | ||
| NM_178780.3 | NP_848895.2 | 2 | 256 | |||
| GenBank mRNA | AK038724.1 | - | 2 | 255 | ||
| AK053760.1 | - | 2 | 237 | |||
| AK082275.1 | - | 2 | 237 | |||
| AK134663.1 | - | 2 | 238 | |||
| AK138641.1 | - | 2 | 255 | |||
| AK149334.1 | - | 2 | 244 | |||
| BY241896.1 | - | 2 | 263 | |||
| BY279630.1 | - | 2 | 258 | |||

Ric3
Genomic Map