siRNA Details
| Gene Symbol: Klri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177155.4 | NP_796129.2 | 2 | 311 | ||
| GenBank mRNA | AK040915.1 | - | 2 | 298 | ||
| AK041213.1 | - | 2 | 314 | |||
| AY324873.1 | - | 2 | 264 | |||
| BC132406.1 | - | 2 | 291 | 40130683 | ||
| BC132408.1 | - | 2 | 291 | 40130685 | ||

Klri2
Genomic Map