siRNA Details
Gene Symbol: Trank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164659.1 | NP_001158131.1 | 10 | 1852 | ||
GenBank mRNA | AK079552.1 | - | 10 | 1853 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164659.1 | NP_001158131.1 | 10 | 1852 | ||
GenBank mRNA | AK079552.1 | - | 10 | 1853 |