siRNA Details
| Gene Symbol: Trank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164659.1 | NP_001158131.1 | 11 | 4083 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trank1| Gene Symbol: Trank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164659.1 | NP_001158131.1 | 11 | 4083 | ||