siRNA Details
| Gene Symbol: Alg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081264.1 | NP_001074733.1 | 13, 14 | 1326 | ||
| GenBank mRNA | AK157903.1 | - | 13, 14 | 1288 | ||
| AK171887.1 | - | 13, 14 | 1279 | |||
| BC044875.1 | - | 3, 4 | 161 | |||
| BC050854.1 | - | 13, 14 | 1291 | 6332345 | ||
| BC080867.1 | - | 12, 13 | 3283 | |||

Alg6
Genomic Map