siRNA Details
| Gene Symbol: Tmem215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166009.1 | NP_001159481.1 | 2 | 1014 | ||
| NM_177175.4 | NP_796149.2 | 2 | 1956 | |||
| GenBank mRNA | AK044217.1 | - | 2 | 1958 | ||
| AK044627.1 | - | 2 | 1958 | |||
| BC144984.1 | - | 2 | 1011 | 9053532 | ||
| BC148227.1 | - | 2 | 1088 | 40140720 | ||
| BC148284.1 | - | 2 | 1088 | 40140721 | ||

Tmem215
Genomic Map