siRNA Details
Gene Symbol: Vps13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177184.3 | NP_796158.2 | 11, 12 | 853 | ||
GenBank mRNA | AB281675.1 | - | 11, 12 | 853 | ||
AK048766.1 | - | 11, 12 | 838 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vps13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177184.3 | NP_796158.2 | 11, 12 | 853 | ||
GenBank mRNA | AB281675.1 | - | 11, 12 | 853 | ||
AK048766.1 | - | 11, 12 | 838 |