siRNA Details
| Gene Symbol: Svopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177200.4 | NP_796174.2 | 15, 16 | 1575 | ||
| GenBank mRNA | AK078436.1 | - | 5, 6 | 545 | ||
| BC058741.1 | - | 15, 16 | 1558 | 6309979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Svopl| Gene Symbol: Svopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177200.4 | NP_796174.2 | 15, 16 | 1575 | ||
| GenBank mRNA | AK078436.1 | - | 5, 6 | 545 | ||
| BC058741.1 | - | 15, 16 | 1558 | 6309979 | ||