siRNA Details
| Gene Symbol: Dopey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177208.3 | NP_796182.2 | 21 | 3448 | ||
| GenBank mRNA | AK045873.1 | - | 20 | 3329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dopey1| Gene Symbol: Dopey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177208.3 | NP_796182.2 | 21 | 3448 | ||
| GenBank mRNA | AK045873.1 | - | 20 | 3329 | ||