siRNA Details
| Gene Symbol: Dopey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177208.3 | NP_796182.2 | 27 | 6013 | ||
| GenBank mRNA | AK038064.1 | - | 4 | 1089 | ||
| AK047998.1 | - | 5 | 518 | |||
| BC043710.1 | - | 2 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dopey1| Gene Symbol: Dopey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177208.3 | NP_796182.2 | 27 | 6013 | ||
| GenBank mRNA | AK038064.1 | - | 4 | 1089 | ||
| AK047998.1 | - | 5 | 518 | |||
| BC043710.1 | - | 2 | 234 | |||