siRNA Details
| Gene Symbol: Snrnp200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177214.4 | NP_796188.2 | 23, 24 | 3267 | ||
| GenBank mRNA | AK173025.1 | - | 18, 19 | 2496 | ||
| BC063261.1 | - | 23, 24 | 3267 | 6408607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrnp200| Gene Symbol: Snrnp200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177214.4 | NP_796188.2 | 23, 24 | 3267 | ||
| GenBank mRNA | AK173025.1 | - | 18, 19 | 2496 | ||
| BC063261.1 | - | 23, 24 | 3267 | 6408607 | ||