siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161515.1 | NP_001154987.1 | 4 | 629 | ||
| NM_001161516.1 | NP_001154988.1 | 4 | 410 | |||
| NM_178788.4 | NP_848903.2 | 4 | 651 | |||
| GenBank mRNA | AK031645.1 | - | 4 | 616 | ||
| AK039066.1 | - | 4 | 441 | |||
| AK077663.1 | - | 4 | 613 | |||
| AK138614.1 | - | 4 | 439 | |||
| AK171894.1 | - | 4 | 448 | |||
| BB193146.2 | - | 2 | 220 | |||
| BC031719.1 | - | 4 | 435 | 3672932 | ||
| BQ572929.1 | - | 2 | 125 | |||
| CO803572.1 | - | 4 | 410 | |||

Dctd
Genomic Map