siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161515.1 | NP_001154987.1 | 5 | 716 | ||
| NM_001161516.1 | NP_001154988.1 | 5 | 497 | |||
| NM_178788.4 | NP_848903.2 | 5 | 738 | |||
| GenBank mRNA | AK031645.1 | - | 5 | 703 | ||
| AK039066.1 | - | 5 | 528 | |||
| AK077663.1 | - | 5 | 700 | |||
| AK171894.1 | - | 5 | 1138 | |||
| CO803572.1 | - | 5 | 497 | |||

Dctd
Genomic Map