siRNA Details
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161515.1 | NP_001154987.1 | 6 | 780 | ||
NM_001161516.1 | NP_001154988.1 | 6 | 561 | |||
NM_178788.4 | NP_848903.2 | 6 | 802 | |||
GenBank mRNA | AK031645.1 | - | 6 | 767 | ||
AK039066.1 | - | 6 | 592 | |||
AK077663.1 | - | 6 | 764 | |||
AK171894.1 | - | 6 | 1202 | |||
BC031719.1 | - | 6 | 586 | 3672932 | ||
CO803572.1 | - | 6 | 561 |
