siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177240.3 | NP_796214.2 | 3 | 432 | ||
| GenBank mRNA | AK083445.1 | - | 3 | 429 | ||
| AK085769.1 | - | 3 | 433 | |||
| AK160812.1 | - | 3 | 433 | |||
| BC147504.1 | - | 3 | 350 | 9056419 | ||
| BC151159.1 | - | 3 | 350 | 9088048 | ||
| BY713400.1 | - | 3 | 433 | |||

Trappc11
Genomic Map