siRNA Details
Gene Symbol: Spata33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177279.4 | NP_796253.2 | 3 | 404 | ||
GenBank mRNA | AK076656.1 | - | 3 | 380 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spata33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177279.4 | NP_796253.2 | 3 | 404 | ||
GenBank mRNA | AK076656.1 | - | 3 | 380 |