siRNA Details
| Gene Symbol: Itgb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177290.3 | NP_796264.2 | 14 | 2766 | ||
| GenBank mRNA | AK140969.1 | - | 5 | 851 | ||
| BC120691.1 | - | 14 | 2766 | 40129614 | ||
| BC125343.1 | - | 14 | 2766 | 40129858 | ||
| BC144785.1 | - | 14 | 2771 | |||

Itgb8
Genomic Map