siRNA Details
| Gene Symbol: Serac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111017.1 | NP_001104487.1 | 12 | 1332 | ||
| NM_177311.4 | NP_796285.2 | 11 | 1242 | |||
| GenBank mRNA | AK019677.1 | - | 11 | 1231 | ||
| AK154605.1 | - | 12 | 1283 | |||
| AK155571.1 | - | 12 | 1335 | |||
| AK170498.1 | - | 12 | 1283 | |||
| BC119594.1 | - | 12 | 1321 | 40059066 | ||
| BC119595.1 | - | 12 | 1321 | 40059071 | ||

Serac1
Genomic Map