siRNA Details
| Gene Symbol: E130307A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049799.1 | - | 1 | 41 | ||
| AK164126.1 | - | 1 | 41 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130307A14Rik| Gene Symbol: E130307A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049799.1 | - | 1 | 41 | ||
| AK164126.1 | - | 1 | 41 | |||