siRNA Details
Gene Symbol: Ubtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173784.3 | NP_776145.2 | 2 | 385 | ||
GenBank mRNA | AK076953.1 | - | 2 | 387 | ||
BC057026.1 | - | 2 | 385 | 6825513 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173784.3 | NP_776145.2 | 2 | 385 | ||
GenBank mRNA | AK076953.1 | - | 2 | 387 | ||
BC057026.1 | - | 2 | 385 | 6825513 |