siRNA Details
| Gene Symbol: Spata32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177801.3 | NP_808469.1 | 4 | 767 | ||
| GenBank mRNA | AK077068.1 | - | 4 | 768 | ||
| BC117072.1 | - | 4 | 767 | 40126391 | ||
| BC120775.1 | - | 4 | 766 | 40129698 | ||
| DQ118647.1 | - | 4 | 753 | |||

Spata32
Genomic Map