siRNA Details
| Gene Symbol: Fads6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178035.4 | NP_828874.3 | 6 | 989 | ||
| GenBank mRNA | AB087503.1 | - | 6 | 957 | ||
| BC044804.1 | - | 7 | 797 | |||
| BC132041.1 | - | 6 | 940 | 40130318 | ||
| BC132067.1 | - | 6 | 940 | 40130344 | ||

Fads6
Genomic Map