siRNA Details
| Gene Symbol: Gm9776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041197.1 | - | 1 | 190 | ||
| AK141947.1 | - | 109 | ||||
| AK162882.1 | - | 1 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9776| Gene Symbol: Gm9776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041197.1 | - | 1 | 190 | ||
| AK141947.1 | - | 109 | ||||
| AK162882.1 | - | 1 | 216 | |||