siRNA Details
Gene Symbol: Gm9776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041197.1 | - | 1 | 395 | ||
AK141947.1 | - | 314 | ||||
AK162882.1 | - | 1 | 421 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041197.1 | - | 1 | 395 | ||
AK141947.1 | - | 314 | ||||
AK162882.1 | - | 1 | 421 |