siRNA Details
| Gene Symbol: Zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183208.3 | NP_899031.2 | 16 | 2497 | ||
| GenBank mRNA | BC057691.1 | - | 6 | 734 | ||
| BC058646.1 | - | 16 | 2397 | 6405777 | ||
| BC065120.1 | - | 16 | 2390 | 6856060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmiz1| Gene Symbol: Zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183208.3 | NP_899031.2 | 16 | 2497 | ||
| GenBank mRNA | BC057691.1 | - | 6 | 734 | ||
| BC058646.1 | - | 16 | 2397 | 6405777 | ||
| BC065120.1 | - | 16 | 2390 | 6856060 | ||