siRNA Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181345.3 | NP_851990.2 | 5, 6 | 531 | ||
| GenBank mRNA | AK054533.1 | - | 5, 6 | 532 | ||
| AK139497.1 | - | 5, 6 | 532 | |||
| AY262112.1 | - | 5, 6 | 531 | |||
| BC048751.1 | - | 5, 6 | 506 | |||
| BC104058.1 | - | 5, 6 | 493 | 40040361 | ||
| BC104059.1 | - | 5, 6 | 493 | 40040362 | ||

Npm2
Genomic Map