siRNA Details
| Gene Symbol: D630010B17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085312.1 | - | 4 | 821 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630010B17Rik| Gene Symbol: D630010B17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085312.1 | - | 4 | 821 | ||