siRNA Details
| Gene Symbol: Vwa5b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144953.1 | NP_001138425.1 | 22 | 3488 | ||
| GenBank mRNA | AK045595.1 | - | 1 | 1478 | ||
| BC110635.2 | - | 1 | 1217 | |||
| BC125016.1 | - | 1 | 1219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa5b2| Gene Symbol: Vwa5b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144953.1 | NP_001138425.1 | 22 | 3488 | ||
| GenBank mRNA | AK045595.1 | - | 1 | 1478 | ||
| BC110635.2 | - | 1 | 1217 | |||
| BC125016.1 | - | 1 | 1219 | |||