siRNA Details
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177375.1 | NP_796349.1 | 2 | 445 | ||
GenBank mRNA | AB232621.2 | - | 2 | 281 | ||
AK043912.1 | - | 2 | 410 | |||
AK080607.1 | - | 425 | ||||
BC094931.1 | - | 2 | 353 | 6436242 | ||
BC118018.1 | - | 2 | 586 | 40103722 |
