siRNA Details
| Gene Symbol: Clpsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034871.2 | NP_001030043.1 | 1 | 54 | ||
| GenBank mRNA | AK136682.1 | - | 1 | 56 | ||
| BC147116.1 | - | 1 | 130 | 9056160 | ||
| BC147117.1 | - | 1 | 130 | 9056121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clpsl2| Gene Symbol: Clpsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034871.2 | NP_001030043.1 | 1 | 54 | ||
| GenBank mRNA | AK136682.1 | - | 1 | 56 | ||
| BC147116.1 | - | 1 | 130 | 9056160 | ||
| BC147117.1 | - | 1 | 130 | 9056121 | ||