siRNA Details
| Gene Symbol: Zscan30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049082.1 | - | 3 | 504 | ||
| BC117067.1 | - | 2 | 285 | 40126386 | ||
| BC117069.1 | - | 2 | 285 | 40126388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zscan30| Gene Symbol: Zscan30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049082.1 | - | 3 | 504 | ||
| BC117067.1 | - | 2 | 285 | 40126386 | ||
| BC117069.1 | - | 2 | 285 | 40126388 | ||