siRNA Details
Gene Symbol: Scd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183216.3 | NP_899039.2 | 2 | 283 | ||
GenBank mRNA | AY430080.1 | - | 2 | 283 | ||
BC141234.1 | - | 2 | 239 | 9055846 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183216.3 | NP_899039.2 | 2 | 283 | ||
GenBank mRNA | AY430080.1 | - | 2 | 283 | ||
BC141234.1 | - | 2 | 239 | 9055846 |