siRNA Details
| Gene Symbol: 9130227L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033706.1 | - | 3 | 535 | ||
| AK079812.1 | - | 6 | 1033 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130227L01Rik| Gene Symbol: 9130227L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033706.1 | - | 3 | 535 | ||
| AK079812.1 | - | 6 | 1033 | |||