siRNA Details
| Gene Symbol: Fam163a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177838.3 | NP_808506.1 | 884 | |||
| GenBank mRNA | AB299026.1 | - | 2 | 440 | ||
| AK039197.1 | - | 885 | ||||
| BC116972.1 | - | 2 | 553 | 40126291 | ||
| BC116974.1 | - | 2 | 553 | 40126293 | ||

Fam163a
Genomic Map