siRNA Details
| Gene Symbol: Rnf224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033410.2 | NP_001028582.1 | 2 | 436 | ||
| GenBank mRNA | AK146725.1 | - | 2 | 437 | ||
| BC147425.1 | - | 1 | 629 | 9056501 | ||
| BC147426.1 | - | 1 | 629 | 9056480 | ||

Rnf224
Genomic Map