siRNA Details
| Gene Symbol: Myo3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177376.4 | NP_796350.2 | 20 | 2318 | ||
| GenBank mRNA | AY830392.2 | - | 19 | 2289 | ||
| AY830393.2 | - | 19 | 2289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo3b| Gene Symbol: Myo3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177376.4 | NP_796350.2 | 20 | 2318 | ||
| GenBank mRNA | AY830392.2 | - | 19 | 2289 | ||
| AY830393.2 | - | 19 | 2289 | |||